Skip to Content
Merck
All Photos(1)

Documents

34132

Supelco

3-Acetyldeoxynivalenol solution

~100 μg/mL in acetonitrile, analytical standard

Synonym(s):

3α-Acetoxy-7α,15-dihydroxy-12,13-epoxytrichothec-9-en-8-one, 3α-Acetylvomitoxin, 3-ADON, 3-AcDON

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22O7
CAS Number:
Molecular Weight:
338.35
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CC(=O)O[C@@H]1C[C@@]2(C)[C@]3(CO3)[C@@H]1O[C@@H]4C=C(C)C(=O)[C@@H](O)[C@]24CO

InChI

1S/C17H22O7/c1-8-4-11-16(6-18,13(21)12(8)20)15(3)5-10(23-9(2)19)14(24-11)17(15)7-22-17/h4,10-11,13-14,18,21H,5-7H2,1-3H3/t10-,11-,13-,14-,15-,16-,17+/m1/s1

InChI key

ADFIQZBYNGPCGY-HTJQZXIKSA-N

General description

3-Acetyldeoxynivalenol is a group B trichothecene mycotoxin mainly produced by molds of the genus Fusarium, which is commonly present in feed, especially in cereals and silages.

Application

3-Acetyldeoxynivalenol solution may be used as an analytical reference standard for the quantification of the analyte in bovine milk using liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥97.0% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Bretz et al.
Molecular nutrition & food research, 50(3), 251-260 (2006-03-08)
Trichothecenes are secondary metabolites produced by several fungi of the Fusarium genus during their growth period. They inhibit protein biosynthesis in eukaryotic cells resulting in numerous toxic effects such as diarrhea, vomiting, and gastro-intestinal inflammation. Considering its occurrence in food
Ariadni Geballa-Koukoula et al.
Sensors (Basel, Switzerland), 21(5) (2021-04-04)
In current food safety monitoring, lateral flow immunoassays (LFIAs) are widely used for rapid food contaminant screening. Recent advances include smartphone readouts, offering semi-quantitative analysis of LFIAs with time, location, and data transfer in case of on-site testing. Following the
J J Mateo et al.
Journal of chromatography. A, 918(1), 99-112 (2001-06-14)
Various analytical methods used in the analysis of type B trichothecenes (deoxynivalenol, nivalenol, 3- and 15-acetyldeoxynivalenol) in cereals were compared and optimised in this work. These methods use either GC-electron-capture detection (ECD) of trimethylsilyl, trifluoroacetyl and heptafluorobutyryl derivatives or HPLC
G S Eriksen et al.
Archiv fur Tierernahrung, 57(5), 335-345 (2003-11-19)
The absorption, metabolism and excretion of 3-acetyldeoxynivalenol (3-aDON) in pigs were studied. Pigs with a faecal microflora known to be able to de-epoxidate trichothecenes were used in the experiment. The pigs were fed a commercial diet with 3-aDON added in
A Llorens et al.
International journal of food microbiology, 94(1), 43-54 (2004-06-03)
Various species of Fusarium can produce trichothecene mycotoxins that contaminate food commodities and can represent a risk for human and animal health. In this paper, a full factorial design was applied to study the influence of incubation temperature, water activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service