Skip to Content
Merck
All Photos(1)

Key Documents

442734

Supelco

p-Terphenyl-d14

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5C6D4C6D5
CAS Number:
Molecular Weight:
244.39
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

212-213 °C (lit.)

application(s)

environmental

format

neat

mass shift

M+14

storage temp.

2-30°C

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])-c2c([2H])c([2H])c(c([2H])c2[2H])-c3c([2H])c([2H])c([2H])c([2H])c3[2H]

InChI

1S/C18H14/c1-3-7-15(8-4-1)17-11-13-18(14-12-17)16-9-5-2-6-10-16/h1-14H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D

InChI key

XJKSTNDFUHDPQJ-WZAAGXFHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Praew Suppajariyawat et al.
Forensic science international, 301, 415-425 (2019-06-23)
Ammonium nitrate fuel oil (ANFO) is one of the most favorite explosives used in terrorist attacks. This explosive is a complex mixture of 95-96% ammonium nitrate (AN) and 4-5% liquid hydrocarbons (fuel oil). In this study, we analyze a variety

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service