Skip to Content
Merck
All Photos(1)

Key Documents

364630

Sigma-Aldrich

p-Terphenyl-d14

98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€53.60
500 MG
€173.00

€53.60


Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
€53.60
500 MG
€173.00

About This Item

Linear Formula:
C6D5C6D4C6D5
CAS Number:
Molecular Weight:
244.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12

€53.60


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

mp

212-213 °C (lit.)

application(s)

electronics

mass shift

M+14

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])-c2c([2H])c([2H])c(c([2H])c2[2H])-c3c([2H])c([2H])c([2H])c([2H])c3[2H]

InChI

1S/C18H14/c1-3-7-15(8-4-1)17-11-13-18(14-12-17)16-9-5-2-6-10-16/h1-14H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D

InChI key

XJKSTNDFUHDPQJ-WZAAGXFHSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

May be used as an analytical standard

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victoria A Sannikova et al.
The journal of physical chemistry. A, 123(2), 505-516 (2018-12-20)
Magnetic field-affected reaction yield (MARY) spectroscopy is a spin chemistry technique for detecting short-lived radical ions. Having sensitivity to transient species with lifetimes as short as nanoseconds, MARY spectroscopy usually does not provide detailed information on their magnetic resonance parameters
Noriatsu Ozaki et al.
Water research, 80, 337-345 (2015-06-01)
The loading and removal of polycyclic aromatic hydrocarbons (PAHs) were measured and estimated in a wastewater treatment plant in a separated sewer system in a suburban area of Japan. The influent 16 PAHs concentration was 219 ± 210 ng L(-1)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service