Skip to Content
Merck
All Photos(1)

Key Documents

442271

Supelco

1-Naphthylamine

analytical standard

Synonym(s):

α-Naphthylamine, 1-Aminonaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NH2
CAS Number:
Molecular Weight:
143.19
Beilstein:
386133
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

301 °C (lit.)

mp

47-50 °C (lit.)

density

1.114 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

room temp

SMILES string

Nc1cccc2ccccc12

InChI

1S/C10H9N/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H,11H2

InChI key

RUFPHBVGCFYCNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaolin Zhang et al.
Inorganic chemistry, 51(4), 2325-2331 (2012-02-10)
A unique gadolinium complex, Nap-DO3A-Gd, comprising a naphthylamine luminescent moiety, a di-2-picolylamine (DPA) binding chelator, and a 1,4,7,10-tetraazacyclododecane-1,4,7-triacetic acid (DO3A) moiety has been designed and synthesized as a dual-functional probe for selective magnetic resonance imaging and fluorescent sensing of copper(II)
Arvydas Tamulis et al.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life, 43(1), 49-66 (2012-12-18)
The quantum mechanical self-assembly of two separate photoactive supramolecular systems with different photosynthetic centers was investigated by means of density functional theory methods. Quantum entangled energy transitions from one subsystem to the other and the assembly of logically controlled artificial
V Chubanov et al.
British journal of pharmacology, 166(4), 1357-1376 (2012-01-17)
Transient receptor potential cation channel subfamily M member 7 (TRPM7) is a bifunctional protein comprising a TRP ion channel segment linked to an α-type protein kinase domain. TRPM7 is essential for proliferation and cell growth. Up-regulation of TRPM7 function is
Silvia Spinelli et al.
Insect biochemistry and molecular biology, 42(1), 41-50 (2011-11-15)
Apis mellifera (Amel) relies on its olfactory system to detect and identify new-sources of floral food. The Odorant-Binding Proteins (OBPs) are the first proteins involved in odorant recognition and interaction, before activation of the olfactory receptors. The Amel genome possess
Yuta Matsuoka et al.
Free radical biology & medicine, 53(11), 2112-2118 (2012-10-03)
Ascorbic acid is a small-molecule reductant with multiple functions in vivo. Reducing ascorbic acid intake leads to a lack of hydroxylation of prolines and lysines, causing a looser triple helix and resulting in scurvy. Ascorbic acid also acts as an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service