Key Documents
Z742064
Sigma-Aldrich
KitAlysis Benchtop Inertion Box
Synonym(s):
Benchtop Inertion Box
Select a Size
1 EA
€984.00
€984.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
€984.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
€984.00
Check Cart for Availability
Request a Bulk Order
General description
Provides an inert environment to run oxygen sensitive cross-coupling reactions in a laboratory fume hood.
Designed to be used with KitAlysis High-Throughput Screening Kits and KitAlysis 24-Well Reaction Block.
Features and Benefits
  • Run 24 micro-scale reactions in parallel under an inert atmosphere.
  • Provides a constant flow of inert gas over working area.
  • Easy attachment of tubing (not included) from inert gas valve on hood or stand-alone tank.
Legal Information
KitAlysis is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
KitAlysis High-Throughput Screening Kits provide solution to efficiently identify or optimize suitable catalytic reaction conditions. Chemist can rapidly run 24 unique micro scale reactions in parallel with tailored conditions.
Tools aid in kit setup for organic chemistry techniques, ensuring ease and success.
PhotoKitAlysis Reaction Kit aids in cross-coupling reactions, offering pre-weighed components for optimization.
Materials Included in your KITALYSIS-24PD-2PK High-Throughput Screening Kit
Protocols
Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.
Materials and procedure Step-by-Step Guide for Photocatalysis High-Throughput Reaction Screening Kit
A step-by-step protocol guide for KitAlysis Suzuki-Miyaura Cross-Coupling Reaction Screening Kit.
See All
Related Content
Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.