Skip to Content
Merck
All Photos(1)

Key Documents

41397

Supelco

Tebutam

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

N-Benzyl-N-isopropylpivalamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€92.40

€92.40


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€92.40

About This Item

Empirical Formula (Hill Notation):
C15H23NO
CAS Number:
Molecular Weight:
233.35
Beilstein:
2838127
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€92.40


Estimated to ship on30 May 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

CC(C)N(Cc1ccccc1)C(=O)C(C)(C)C

InChI

1S/C15H23NO/c1-12(2)16(14(17)15(3,4)5)11-13-9-7-6-8-10-13/h6-10,12H,11H2,1-5H3

InChI key

RJKCKKDSSSRYCB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com

Tebutam is a selective benzamide herbicide, used for the pre-emergence control of annual broad-leaved and grass weeds, including volunteer cereals, in oilseed rape and brassicas, at 2800-3600 g/ha.
According to the Commission Implementing Regulation (EU) No 2076/2002 of 20th November 2002, tebutam is not approved for use in the European Union (EU) as a plant protection agent, following the Regulation (EC) No. 1107/2009. But a default maximum residue limit of 0.01 mg/kg is set for the presence of tebutam in products of plant and animal origin, according to Art 18(1)(b) Reg 396/2005.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Tebutam CRM may also be used as given below:

  • Optimization of sample extraction based on QuEChERS and dispersive-solid-phase extraction (d-SPE) clean-up, for the multi-residue analysis of 120 pesticides, 16 polycyclic aromatic hydrocarbons, and 22 polychlorinated biphenyls by liquid and gas chromatography, coupled with tandem mass spectrometry (MS/MS)
  • Evaluation of 250 pesticide residues extracted by QuEChERS method from 103 processed fruit samples using ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • Determination of 90 pesticides, 16 polycyclic aromatic hydrocarbons, and 22 polychlorinated biphenyls in honey samples by using QuEChERS combined with dispersive solid-phase extraction (SPME) for sample pre-treatment and liquid chromatography-triple-quadrupole tandem mass spectrometry and gas chromatography-ion-trap tandem mass spectrometry for quantitation
  • Screening and quantification of 569 pesticides in honeysuckle samples by a combination of ultrahigh-performance liquid chromatography (UHPLC) and quadrupole-Orbitrap high-resolution mass spectrometry
  • Simultaneous determination of 134 pesticide residues in tea samples using d-SPE extraction with a multi-functional filter (MFF), followed by ultra-high performance liquid chromatography (UPLC) and quadrupole time-of-flight mass spectrometry (TOF-MS)

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service