Skip to Content
Merck
All Photos(1)

Key Documents

82280

Sigma-Aldrich

1,2-Propanediol

puriss. p.a., ACS reagent, ≥99.5% (GC)

Synonym(s):

1,2-PDO, Propylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€57.60
1 L
€145.00

€57.60


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€57.60
1 L
€145.00

About This Item

Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1340498
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

€57.60


Estimated to ship on01 June 2025


Request a Bulk Order

grade

ACS reagent
puriss. p.a.

Quality Level

vapor density

2.62 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

≥99.5% (GC)

form

viscous liquid

autoignition temp.

779 °F

shelf life

3 yr

expl. lim.

12.5 %

impurities

≤0.003% free acid (as CH3COOH)
≤0.1% water

evapn. residue

≤0.005%

ign. residue

<0.005% (as SO4)

refractive index

n20/D 1.432 (lit.)
n20/D 1.433

bp

187 °C (lit.)

mp

−60 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Zn: ≤5 mg/kg

SMILES string

CC(O)CO

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3

InChI key

DNIAPMSPPWPWGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1,2-Propanediol is a 1,2-alkanediol. It can be obtained from glycerol, via hydrogenolysis in the presence of Ru/C and Amberlyst ion-exchange resins.[1]

Application

1,2-Propanediol (propanediol) may be employed as a cryoprotective agent for the preservation (by cooling to sub-zero temperatures) of human embryos[2] and oocytes (human and mouse).[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alexander Eberle et al.
Journal of the American Chemical Society, 140(4), 1327-1336 (2018-01-10)
Supramolecular self-assembly at the solid-solid interface enables the deposition and monolayer formation of insoluble organic semiconductors under ambient conditions. The underlying process, termed as the organic solid-solid wetting deposition (OSWD), generates two-dimensional adsorbates directly from dispersed three-dimensional organic crystals. This
Verya Daeichin et al.
IEEE transactions on ultrasonics, ferroelectrics, and frequency control, 64(3), 555-567 (2017-01-24)
Although high-frequency ultrasound imaging is gaining attention in various applications, hardly any ultrasound contrast agents (UCAs) dedicated to such frequencies (>15 MHz) are available for contrast-enhanced ultrasound (CEUS) imaging. Moreover, the composition of the limited commercially available UCAs for high-frequency
Jeffrey J Kim et al.
Tobacco induced diseases, 15, 14-14 (2017-02-28)
Despite the rising health and safety concerns of e-cigarettes, a universal e-cigarette testing method is still in its early developmental stage. The aim of this study was to develop an e-liquid Reference Material that can be used to improve accuracy
D A Gook et al.
Human reproduction (Oxford, England), 8(7), 1101-1109 (1993-07-01)
Human and mouse oocytes were cryopreserved by a slow freeze, rapid thaw method, using propanediol (PROH) as the cryoprotectant. A simulated cryopreservation was also included in the study to detect the level of damage attributable to the PROH alone. Comparison
Iban Seiliez et al.
Autophagy, 12(2), 343-356 (2016-02-24)
Monitoring autophagic flux in vivo or in organs remains limited and the ideal methods relative to the techniques possible with cell culture may not exist. Recently, a few papers have demonstrated the feasibility of measuring autophagic flux in vivo by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service