Skip to Content
Merck
All Photos(1)

Key Documents

33065-U

Supelco

Sylon CT

for GC derivatization, pkg of 400 mL, 5% dimethyldichlorosilane in toluene

Synonym(s):

Dichlorodimethylsilane solution, Dimethyldichlorosilane solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6Cl2Si
CAS Number:
Molecular Weight:
129.06
Beilstein:
605287
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

for GC derivatization

description

Silyl reagent

packaging

pkg of 400 mL

concentration

5% dimethyldichlorosilane in toluene

SMILES string

C[Si](C)(Cl)Cl

InChI

1S/C2H6Cl2Si/c1-5(2,3)4/h1-2H3

InChI key

LIKFHECYJZWXFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For deactivating glassware.
Treat the glass tubing that you use for columns with Sylon-CT solution (5% dimethyldichlorosilane in toluene). It deactivates the tubing for use up to 350 °C to 400 °C, far exceeding the capabilities of other deactivating agents used at room temperature. Sylon CT also deactivates transfer lines and glass or glass-lined injection port liners. Instructions included.

Application

Sylon CT was used as derivatizing agent, during an experiment to develop sensitive GC method for the identification and determination of long-chain primary alkyl amines.[1] It was also used for treating Erlenmeyer flasks to prevent mycelium from sticking to the glassware during development of techniques in the identification and partial characterization the chaxamycin biosynthesis gene cluster of S. leeuwenhoekii.[2]

Legal Information

Sylon CT is a trademark of Sigma-Aldrich Co. LLC

related product

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1A - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Kusch et al.
Journal of chromatography. A, 1113(1-2), 198-205 (2006-02-18)
Gas chromatography with simultaneous flame-ionization detection (FID) and a nitrogen-phosphorus detection (NPD) as well as gas chromatography-mass spectrometry (GC/MS) has been used to characterize long-chain primary alkyl amines after derivatization with trifluoroacetic anhydride (TFAA). Electron impact ionization- (EI) and negative
[A multichannel microelectrode system for long-term use].
I F Kalmykov et al.
Fiziologicheskii zhurnal imeni I.M. Sechenova, 80(8), 135-138 (1994-08-01)
Teemu Näykki
Analytical and bioanalytical chemistry, 372(7-8), 829-831 (2002-05-16)
The use of silylation of glassware in mercury analytics was investigated. By treating the glassware with 5% (v/v) dichlorodimethylsilane in toluene, the surfaces of the vessels were inactivated. Method development proved that silylation can extend the range of stabile, contamination-free
S C Lung et al.
Chemosphere, 41(12), 1865-1871 (2000-11-04)
The effects of coating materials on polychlorinated biphenyl (PCB) adsorption in aqueous solution were assessed in an attempt to minimize PCB sorption loss during sampling processes. A coating material, which enhances PCB adsorption and allows adsorbed PCBs to be readily
Lucas H da Silva et al.
Gerodontology, 29(2), e1019-e1023 (2012-01-10)
The purpose of this study was to evaluate the flexural strength and Vickers hardness of a microwave energy heat-cured acrylic resin by adding different concentrations of silane surface-treated nanoparticle silica. Acrylic resin specimens with dimensions of 65 × 10 ×

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service