Recommended Products
packaging
pkg of 1 ea
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
application(s)
environmental
industrial hygiene
compatibility
for use with thermal desorption tubes
greener alternative category
General description
The kit includes the following:
- Stainless steel 22s gauge needle with bevel point
- Stainless steel fitting
- Acetal/stainless steel thumbwheel
- 1/4 in. PTFE ferrule (white)
- 1/4 in. M-2 VESPEL™ ferrule (orange)
- 6 mm M-2A VESPEL graphite ferrule (black)
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service