27093-U
Assembled screw cap with hole with PTFE/silicone septum
black phenolic hole cap, thread for 8-425, PTFE/silicone, septum thickness 1.5 mm, for use with 2 mL screw thead vial, 4.6 mm opening, pkg of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PTFE/silicone
black phenolic hole cap
feature
closure type black phenolic hole cap
packaging
pkg of 100 ea
septum thickness
1.5 mm
fitting
thread for 8-425
compatibility
for use with 2 mL screw thead vial, 4.6 mm opening
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Biomedicines, 8(4) (2020-04-12)
The multi-omic integration of microbiota data with metabolomics has gained popularity. This protocol is based on a human multi-omics study, integrating cervicovaginal microbiota, HPV status and neoplasia, with urinary metabolites. Indeed, to understand the biology of the infections and to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service