Key Documents
2631605
Supelco
Inlet Liner, for Packed GC (unpacked)
pkg of 5 ea
About This Item
UNSPSC Code:
12000000
Pricing and availability is not currently available.
description
unpacked
packaging
pkg of 5 ea
L × O.D. × I.D.
112 mm × 6.0 mm × 3.0 mm
General description
Deactivated (silanized) glass inlet liner for packed injection ports.
  • Fits PerkinElmer (AutoSystem)
Choose from one of the most recent versions:
Certificates of Analysis (COA)