Skip to Content
Merck
All Photos(1)

Key Documents

1.04419

Supelco

Mercury(II) chloride

for analysis EMSURE® Reag. Ph Eur,ACS

Synonym(s):

Mercury(II) chloride, Mercury dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€123.00
250 G
€246.00
1 KG
€615.00

€123.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€123.00
250 G
€246.00
1 KG
€615.00

About This Item

Empirical Formula (Hill Notation):
Cl2Hg
CAS Number:
Molecular Weight:
271.50
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-299-8
NACRES:
NA.21
grade:
ACS reagent
form:
solid

€123.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

0.0001 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99.5% (complexometric)
≥99.5-100.5% dry basis (complexometric)

form

solid

reduction residue

≤0.02%

loss

≤1% loss on drying, 24 h in vacuum

pH

3.2 (15 g/L in H2O)

bp

302 °C/1013 hPa (sublimed)

mp

280.7 °C

density

5.44 g/cm3 at 20 °C

bulk density

2000 kg/m3

cation traces

Fe: ≤0.002%
HgCl2:, passes test

storage temp.

2-30°C

SMILES string

[Hg+2].[Cl-].[Cl-]

InChI

1S/2ClH.Hg/h2*1H;/q;;+2/p-2

InChI key

LWJROJCJINYWOX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay (complexometric): ≥ 99.5 %
Assay (complexometric; calculated on dried substance): 99.5 - 100.5 %
Identity: passes test
Appearance of solution: passes test
Solution in Ether: passes test
Acidity or alkalinity: passes test
Mercury(I)-chloride: passes test
Fe (Iron): ≤ 0.002 %
Residue after reduction: ≤ 0.02 %
Loss on drying (24 h im Vakuum): ≤ 1.0 %
Correspons to Reag. Ph Eur,ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service