Skip to Content
Merck
All Photos(1)

Key Documents

15022AST

Supelco

Astec® CHIROBIOTIC® V2 Chiral (5 μm) HPLC Columns

L × I.D. 10 cm × 4.6 mm, HPLC Column

Synonym(s):

CHIROBIOTIC V2 Chiral Chromatography Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

Astec® CHIROBIOTIC® V2 Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm

material

stainless steel column

Agency

suitable for USP L88

description

HPLC column

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-45 °C temperature
241 bar pressure (3500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

10 cm × 4.6 mm

matrix

High-purity silica gel particle platform
fully porous particle

matrix active group

vancomycin phase

particle size

5 μm

pore size

200 Å

operating pH range

3.5-7.0

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

Neutral molecules, amides, acids, esters and amines show considerable enantioselectivity on these vancomycin-based CSPs. A wide variety of secondary and tertiary amines have been separated on the CHIROBIOTIC® V in the polar ionic mode. The CHIROBIOTIC® V has demonstrated many of the separation characteristics of protein-based stationary phases, but with exceptional stability and much higher sample capacity. Some chiral analytes have been resolved that have not been reported separated on any other chiral stationary phase. CHIROBIOTIC® V and V2 differ in their bonding chemistry the pore size of the support particle, giving them different selectivity and preparative capacity.

  • Bonded phase: Vancomycin
  • Operating pH range: 3.5 - 7.0
  • Particle diameter: 5, 10 or 16 μm
  • Pore size: 100 Å (CHIROBIOTIC® V) or 200 Å (CHIROBIOTIC® V2)

CHIROBIOTIC FAQs
CHIROBIOTIC Reference Bibliography
Chiral Product Literature

Application


  • Chiral analysis of dextromethorphan and levomethorphan in human hair by liquid chromatography-tandem mass spectrometry: This study utilized the CHIROBIOTIC® V2 column to achieve chiral separation of dextromethorphan and levomethorphan in human hair, highlighting the column′s application in forensic toxicology for precise enantiomeric analysis (Ji et al., 2022).

  • Development and validation of an UFLC-MS/MS method for enantioselectivity determination of d,l-thero-methylphenidate, d,l-thero-ethylphenidate and d,l-thero-ritalinic acid in rat plasma: This research demonstrates the application of CHIROBIOTIC® V2 for pharmacokinetic studies in rats, establishing a method for enantioselective determination that can be crucial for drug development and therapeutic monitoring (Zhang et al., 2016).

  • New high-performance liquid chromatography method for the determination of (R)-warfarin and (S)-warfarin using chiral separation on a glycopeptide-based stationary phase: The use of CHIROBIOTIC® V2 column in developing a new HPLC method for the separation of warfarin enantiomers underscores its utility in clinical analysis and therapeutic drug monitoring, showing its broad applicability in biomedical research (Malakova et al., 2009).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatography Liquid Chiral Separations
Xiao, T.L., et al.
Encyclopedia of Separation Science, 1-15 (2000)
Chiral derivatives of Butenafine and Terbinafine: synthesis and antifungal activity
Fuglseth, Erik, et al.
Tetrahedron, 65 (47), 9807-9813 (2009)
Enantioenriched 1-aryl-2-fluoroethylamines. Efficient lipase-catalysed resolution and limitations to the Mitsunobu inversion protocol
Thvedt, Thor H?kon Krane, et al.
Tetrahedron, 66 (34), 6733-6743 (2010)
Direct and indirect separations of five isomers of Brivanib Alaninate using chiral high-performance liquid chromatography
He, Brian Lingfeng, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 875 (1), 122-135 (2008)
Enantiospecific determination of dl-methylphenidate and dl-ethylphenidate in plasma by liquid chromatography?tandem mass spectrometry: Application to human ethanol interactions
Zhu, Hao-Jie, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 879 (11-12), 783-788 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service