Skip to Content
Merck
All Photos(1)

Documents

10305AST

Supelco

Astec® CHIROBIOTIC® (5 μm) HPLC Column Screening Kit

L × I.D. 25 cm × 4.6 mm

Synonym(s):

Chiral Column Selection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

Astec® CHIROBIOTIC® HPLC Column Screening Kit, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

material

stainless steel column

Quality Level

product line

Astec®

packaging

pkg of 1 kit

manufacturer/tradename

Astec®

parameter

0-45 °C temperature
241 bar pressure (3500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

25 cm × 4.6 mm

matrix

fully porous particle

particle size

5 μm

pore size

100 Å pore size

operating pH

3.5-6.8

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 15024ASTAstec® CHIROBIOTIC® V2 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mmSDS

  • 12024ASTAstec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mmSDS

  • 13024ASTAstec® CHIROBIOTIC® R Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mmSDS

  • 14024ASTAstec® CHIROBIOTIC® TAG Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mmSDS


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatographic Separations and Analysis: Macrocyclic Glycopeptide Chiral Stationary Phases
Berthod, A.
Comprehensive Chirality, 8, 227-262 (2012)
Chromatography Liquid Chiral Separations
Xiao, T.L., et al.
Encyclopedia of Separation Science, 1-15 (2000)
Ying Liu et al.
Journal of chromatography. A, 978(1-2), 185-204 (2002-12-03)
The chiral recognition capabilities of three macrocyclic glycopeptide chiral selectors, namely teicoplanin (Chirobiotic T), its aglycone (Chirobiotic TAG) and ristocetin (Chirobiotic R), were evaluated with supercritical and subcritical fluid mobile phases. A set of 111 chiral compounds including heterocycles, analgesics
Carla Fernandes et al.
Journal of chromatography. A, 1241, 60-68 (2012-05-04)
A chiral HPLC method using four macrocyclic antibiotic chiral stationary phases (CSPs) has been investigated for determination of the enantiomeric purity of fourteen new chiral derivatives of xanthones (CDXs). The separations were performed with the CSPs Chirobiotic T, Chirobiotic TAG
Róbert Berkecz et al.
Journal of chromatography. A, 1216(6), 927-932 (2008-12-27)
The direct separation of the enantiomers of four 2-aminomono- or dihydroxycyclopentanecarboxylic acids and four 2-aminodihydroxycyclohexanecarboxylic acids was performed on chiral stationary phases containing macrocyclic glycopeptide antibiotics such as teicoplanin (Astec Chirobiotic T and T2), teicoplanin aglycone (Chirobiotic TAG) or ristocetin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service