Skip to Content
Merck
All Photos(8)

Documents

ZRB2123

Sigma-Aldrich

Anti-Bacteriophage MS2 Antibody, clone 1B21 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B21, recombinant monoclonal

description

1B21 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 13.86 kDa
observed mol wt ~15 kDa

purified by

using Protein A

species reactivity

virus

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B21 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Capsid protein in Bacteriophage MS2.

Immunogen

His-tagged full-length recombinant Bacteriophage MS2.

Application

Quality Control Testing

Evaluated by Western Blotting in lysates from K562 cells spiked with Bacteriophage MS2.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected Bacteriophage MS2 in lysates from K562 cells spiked with MS2, but not in unspiked lysates.

Tested applications

Affinity Binding Assay: A representative lot of this antibody bound Bacteriophage MS2 with a KD of 3.2 x 10-7 in an affinity binding assay.

ELISA Analysis: 3 μg/mL from a representative lot detected Bacteriophage MS2 in ELISA.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Bacteriophage MS2, clone 1B21 ZooMAb®, Cat. No. ZRB2123, is a recombinant Rabbit monoclonal antibody that detects Bacteriophage MS2 and is tested for use in Affinity Binding Assay, ELISA, and Western Blotting.

Target description

Capsid protein (UniProt: P03612; also known as CP, Coat protein) is encoded by the Cp gene (Gene ID; 1260899) in Enterobacteria phage MS2. MS2 is a small RNA bacteriophage that infects E. coli and other members of the Enterobacteriaceae family. Its genome consists of a single sense (+) strand of RNA 3569 nucleotides in length, which encodes only four proteins: the coat protein, the A or maturation protein that is required for infectivity, a subunit of the viral replicase, and a small lysis protein encoded in an overlapping reading frame. The capsid protein acts as a translational repressor of viral replicase synthesis late in infection. This latter function is the result of capsid protein interaction with an RNA hairpin which contains the replicase ribosome-binding site. Capsid protein self-assembles to form an icosahedral capsid with a T=3 symmetry that is about 26 nm in diameter and consists of 89 capsid proteins dimers (178 capsid proteins). Capsid protein is shown to be a homodimer that can form dimers that assemble by group of 5. Twelve such pentamers are linked together with free dimers. The homodimers bind to the viral RNA via an operator hairpin. It also binds to many other RNA sequences in the viral genome and this interaction probably shifts the virus from the replicative to the assembly phase and ensures specific encapsidation of the viral genome. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Golmohammadi, R., et al. (1993). J. Mol. Biol. 234(3); 620-639).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service