Skip to Content
Merck
All Photos(18)

Key Documents

ZRB1657

Sigma-Aldrich

Anti-phospho-TSC2-Ser939 Antibody, clone 1G19 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Tuberous sclerosis 2 protein

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€395.00

€192.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€192.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G19, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 200.61 kDa
observed mol wt ~220 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
peptide synthesis: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer939)

Gene Information

human ... TSC2(7249)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1G19 is a ZooMAb® Rabbit recombinant monoclonal antibody that detects Tuberin/TSC2. It targets an epitope within 10 amino acids surrounding phosphoserine 939.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphoserine 939 from the internal region of human Tuberin/TSC2.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa treated with Anisomycin.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected phospho-Tuberin/TSC2-Ser939 in lysate from HeLa cells treated with Anisomycin (25 μg/mL; 30 min) but not in untreated cells.

Tested applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected phospho-TSC2-Ser939 in Hek293 treated with Anisomycin and phospho-TSC2/Tuberin-Ser939 Rinehart.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected phospho-TSC2-Ser939 in human kidney and human prostate tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-TSC2-Ser939 in HEK 293 treated with Anisomycin.

Peptide Inhibition Assay: Target band detection in lysate from HeLa cells treated with Anisomycin was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Tuberin (UniProt: P49815; also known as Tuberous sclerosis 2 protein) is encoded by the TSC2 (also known as TSC4) gene (Gene ID: 7249) in human. TSC2 is a peripheral membrane protein that serves as a tumor suppressor and is expressed in multiple tissues. In complex with TSC1, it inhibits the nutrient-mediated or growth factor-stimulated phosphorylation of S6K1 and EIF4EBP1 by negatively regulating mTORC1 signaling. It acts as a GTPase-activating protein (GAP) for the small GTPase Rheb, a direct activator of the protein kinase activity of mTORC1. In its C-terminal region is a highly conserved Rap-Gap domain (aa 1531-1758) that is essential for its function. TSC2 has been reported to increase the intrinsic GTPase activity of both Rap1A and Rab5 in vitro. It can undergo phosphorylation at serine 1387, 1418 or 1420. However, these phosphorylations do not affect its interaction with TSC1. It can also undergo phosphorylation at serine 939 and threonine 1462 by Akt1 induced by growth factor stimulation. When phosphorylated and activated by AMPK under conditions of low ATP:AMP ratio, it leads to negative regulation of the mTORC1 complex. AMPK-mediated phosphorylation leads to TSC1-TSC2 complex stabilization, increasing GAP activity toward Rheb and allows turning off mTORC1 signaling. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Li, Y., et al. (2004). Trends Biochem. Sci. 29(1); 32-38; Tee, AR., et al. (2002). Proc. Natl. Acad. Sci. USA. 99(21); 13571-13576

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service