Skip to Content
Merck

Skip To

T103

Trifluperidol hydrochloride

Synonym(s):

4′-Fluoro-4-[4-hydroxy-4-(α,α,α-trifluoro-m tolyl)piperidino]butyrophenone hydrochloride, R 2498, Triperidol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 327,00

About This Item

Empirical Formula (Hill Notation):
C22H23F4NO2 · HCl
CAS Number:
Molecular Weight:
445.88
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
218-170-1
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
desiccated

€ 327,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to off-white

solubility

DMSO: ≥20 mg/mL, H2O: soluble, ethanol: moderately soluble

SMILES string

Cl.OC1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)c3cccc(c3)C(F)(F)F

InChI

1S/C22H23F4NO2.ClH/c23-19-8-6-16(7-9-19)20(28)5-2-12-27-13-10-21(29,11-14-27)17-3-1-4-18(15-17)22(24,25)26;/h1,3-4,6-9,15,29H,2,5,10-14H2;1H

InChI key

VYGQXRZAHIZHQV-UHFFFAOYSA-N

Gene Information

Application

The interaction of Trifluperidol hydrochloride with NMDA receptors was studied in rat brain buffy coat membranes.[1]

Biochem/physiol Actions

Trifluperidol is a dopamine receptor antagonist with antipsychotic effects used in the treatment of schizophrenia.[2] This neuroleptic penetrates the brain and also affects the glial cells leading to increased release of proinflammatory cytokines.[3] It also alters the metabolism of carbohydrates and amino acids in the brain cortex.[4]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2841A2846N7127
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98%

form

solid

form

powder

form

solid

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

200

solubility

DMSO: ≥20 mg/mL, ethanol: moderately soluble, H2O: soluble

solubility

DMSO: 2 mg/mL, clear

solubility

H2O: soluble

solubility

H2O: 50 mg/mL

storage condition

desiccated

storage condition

-

storage condition

desiccated

storage condition

-

color

white to off-white

color

white to beige

color

off-white to light tan

color

-


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters