Skip to Content
Merck
All Photos(13)

Key Documents

ZRB1285

Sigma-Aldrich

Anti-Thy-1(CD90) Antibody, clone 1I7 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€395.00

€192.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€192.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1I7, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 17.94 kDa
observed mol wt ~24 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1I7 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Thy-1 membrane glycoprotein (CD90). It targets an epitope within 19 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the internal region of human Thy-1 (CD90) membrane glycoprotein.

Application

Quality Control Testing

Evaluated by Western Blotting in SK-N-SH cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Thy-1(CD90) in SK-N-SH cell lysate.

Tested applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected Thy-1(CD90) in K562 and TF-1 cell lysates.

Immunocytochemistry Analysis: A 1:1000 dilution from a representative lot detected Thy-1(CD90) in SK-N-SH cells.

Affinity Binding Assay: A representative lot of this antibody bound THY-1 (CD90) peptide with a KD of 2.1 x 10-7 in an affinity binding assay.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected Thy-1(CD90) in one million TF1 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Thy-1(CD90) in human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Thy-1(CD90), clone 1I7 ZooMAb®, Cat. No. ZRB1285, is a recombinant Rabbit monoclonal antibody that detects Thy-1 (CD90) and is tested in Affinity Binding Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, and Western Blotting.

Target description

Thy-1 membrane glycoprotein (UniProt: P04216; also known as CDw90, Thy-1 antigen, CD90) is encoded by the THY1 gene (Gene ID: 7070) in human. Thy-1 is a highly conserved, GPI-anchored membrane protein with one Ig V-type superfamily domain (aa 20-126). It is synthesized with a signal peptide (aa 1-19) and a propeptide (aa 131-161) that are subsequently cleaved off to generate the mature form (aa 20-130). It lacks an intracellular domain but is located in the outer leaflet of lipid rafts at the cell plasma membrane allowing signaling functions by cis- and trans-interactions with G inhibitory proteins, the Src and c-fyn kinases, and tubulin. Thy-1 is a major cell surface protein found on both T-cells and in the brain. It plays a key role in cell-cell and cell-ligand interactions during synaptogenesis and other events in the brain. It also plays a role in the regulation of axon growth and nerve regeneration, T cell activation and apoptosis, leukocytes and melanoma cell adhesion and migration, and fibroblast proliferation and migration in wound healing. Thy-1 mRNA and protein expression has been reported in several cancer types including liver, myeloid, skin, and brain. It is considered as a marker for glioblastoma stem cells. Reduced expression of Thy-1 is shown to enhance the expression of osteogenic and adipogenic differentiation of mesenchymal stem cells. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Sauzay, C., et al. (2019). Front. Cell Dev. Biol. 7; 66; Moraes, DA., et al. (2016). Stem Cell Res. Ther. 7; Article 97).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service