Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700706

Sigma-Aldrich

Monoclonal Anti-CD90-FITC antibody produced in mouse

clone 5E10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Thy-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 TEST
€341.00

€341.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 TEST
€341.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

€341.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5E10, monoclonal

form

buffered aqueous solution

species reactivity

pig, nonhuman primates, human, dog

technique(s)

flow cytometry: suitable
immunoblotting: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... THY1(7070)

Related Categories

General description

Thy-1 cell surface antigen (Thy-1)/CD90 (cluster of differentiation 90) is a 25–37kDa glycosylphosphatidylinositol (GPI)-anchored glycoprotein. It was first detected in the T cells of mice. Thy-1 is expressed in thymocytes, T cells, neurons, hematopoietic stem cells, cancer stem cells, endothelial cells and fibroblasts. This gene is located on human chromosome 11q23.

Immunogen

HEL erythroleukemia cells

Biochem/physiol Actions

Thy-1 cell surface antigen (Thy-1)/CD90 (cluster of differentiation 90) is involved in T-cell activation, neuritis outgrowth modulation, vesicular release of neurotransmitter at the synapse, astrocyte adhesion, apoptosis in carcinoma cells, tumor suppression, wound healing, fibrosis and fibrogenesis. It also controls fibroblast focal adhesion, cytoskeleton organization and cell migration.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A reduction in CD90 (THY-1) expression results in increased differentiation of mesenchymal stromal cells.
Moraes DA, et al.
Stem Cell Research & Therapy, 7(1), 97-97 (2016)
Further mapping of an ataxia-telangiectasia locus to the chromosome 11q23 region.
Sanal O, et al.
American Journal of Human Genetics, 47(5), 860-860 (1990)

Articles

PLTMax® Human Platelet Lysate (hPL) is a superior serum-free and xeno-free media supplement alternative to fetal bovine serum (FBS) for human mesenchymal stem cell (MSC) cultures.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service