Skip to Content
Merck
All Photos(15)

Key Documents

ZRB1268

Sigma-Aldrich

Anti-Prion Protein Antibody, clone 3D17 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€163.00
4 X 25 μL
€336.00

€163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€163.00
4 X 25 μL
€336.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€163.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3D17, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~27.66 kDa
observed mol wt ~35 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3D17 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Prion protein. It targets an epitope within 19 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal half of human Prion protein.

Application

Quality Control Testing

Evaluated by Western Blotting in Human brain tissue lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Prion Protein in Human brain tissue lysate.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Prion Protein in human cerebral cortex and human Alzheimer′s diseased brain tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound Prion peptides with a KD of 8.4 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Prion Protein in SH-SY5Y cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Prion Protein, clone 3D17 ZooMAb®, Cat. No. ZRB1268, is a recombinant Rabbit monoclonal antibody that specifically targets Prion Protein and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry, and Western Blotting

Target description

Major prion protein (UniProt: P04156; also known as PrP, ASCR, PrP27-30, PrP33-35C, CD230) is encoded by the PRNP (also known as ALTPRP, PRIP, PRP) gene (Gene ID: 5621) in human. PrP is highly expressed in the brain, lung, kidney and heart and low levels of expression are reported in the liver and spleen. It is found in high quantity in the brain of humans and animals infected with degenerative neurological diseases such as Kuru, Creutzfeldt-Jakob disease (CJD), Gerstmann-Straussler syndrome (GSS), scrapie, bovine spongiform encephalopathy (BSE), and transmissible mink encephalopathy (TME). In normal brain it may play a role in neuronal development and synaptic plasticity and may be required for neuronal myelin sheath maintenance and myelin homeostasis through acting as an agonist for ADGRG6 receptor. PrP can exist in either PrPc (c = cellular) or PrPsc (sc = Scrapie) form. PrPc is a glycoprotein normally found inserted in the plasma membrane by a glycosyl phosphatidylinositol (GPI) anchor and can be easily digested by proteases. It is composed mainly of -helices. The PrPsc has more of a -sheet type structure and is highly resistant to the action of proteases. The prion proteins possess two N-linked glycosylation sites and may contain over 50 different sugars. The large size of the N-linked sugars enables them to shield two orthogonal faces of the protein almost completely and protect large regions of the protein surface from proteases. The PrPsc is shown to contain the same set of glycans as PrPc but has a higher proportion of tri- and tetra-antennary sugars. It is shown that following proteinase K treatment, PrPc is completely digested, whereas PrPsc is only shortened to a 27-30 kDa fragment. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service