Skip to Content
Merck
All Photos(18)

Key Documents

ZRB1059

Sigma-Aldrich

Anti-PSMB2 Antibody, clone 2N18 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:3.4.25.1, Macropain subunit C7-I, Multicatalytic endopeptidase complex subunit C7-I, Proteasome component C7-I, Proteasome subunit beta type-2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€163.00
4 X 25 μL
€336.00

€163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€163.00
4 X 25 μL
€336.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€163.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2N18, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 22.84 kDa
observed mol wt ~19 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PSMB2(5690)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 2N18 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human Proteasome subunit beta type-2 (PSMB2).

Immunogen

His-tagged full-length recombinant human Proteasome subunit beta type-2 (PSMB2).

Application

Anti-PSMB2, clone 2N18 ZooMAb, Cat. No. ZRB1059, is a recombinant Rabbit monoclonal antibody that specifically targets PSMB2 and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected PSMB2 in Jurkat and K562 cell lysates.

Affinity Binding Assay: A representative lot of this antibody bound recombinant PSMB2 protein with a KD of 3.1 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected PSMB2 in human small intestine tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PSMB2 in HeLa, HUVEC, and A431 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Proteasome subunit b type-2 (P49721; also known as EC:3.4.25.1, Macropain subunit C7-I, Multicatalytic endopeptidase complex subunit C7-I, Proteasome component C7-I) is encoded by the PSMB2 gene (Gene ID: 5690) in human. Proteasomes are large multi-subunit protease complexes, localized in the nucleus and cytosol that selectively degrade intracellular proteins. Proteasome consists of a multi-component enzymatic complex known as the 26S proteasome, which plays a key role in the maintenance of protein homeostasis by removing misfolded or damaged proteins that could impair cellular functions, and by removing proteins whose functions are no longer required. The proteolytic core of this complex, the 20S proteasome, contains multiple peptidase activities and functions as the catalytic machine. Unlike other proteases, the 20S proteasome ensures that virtually all peptide bonds within a protein substrate are susceptible to cleavage, by possessing multiple proteolytic activities in one proteolytic internal chamber. This core is composed of 28 subunits arranged in four heptameric, tightly stacked, rings ( 7, β7, β7, 7) to form a cylindrical structure. The proteolytic activity is confined to the β1, β2, and β5-subunits. The proteolytic sites are formed by N-terminal threonine residues that face the central cavity of the 20S complex. The regulatory unit of the 26S proteasome is known as the 19S (PA 700) particle and consists of at least 15 subunits that include ATPases, a de-ubiquitinating enzyme, and polyubiquitin-binding subunits. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service