Skip to Content
Merck
All Photos(11)

Key Documents

ZMS1030

Sigma-Aldrich

Anti-Integrin αVβ3 Antibody, clone LM609 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CD51/CD61

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€192.00
4 X 25 μL
€395.00

€192.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€192.00
4 X 25 μL
€395.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€192.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

LM609, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 116.04 kDa

species reactivity

human

species reactivity (predicted by homology)

porcine, avian, bovine, rabbit, monkey, canine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)
pig ... Itgav(397285)
rhesus monkey ... Itgav(707756)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone LM609 is a ZooMAb mouse recombinant monoclonal antibody that specifically detects Integrin aVβ3 .

Immunogen

Full length purified Integrin aV β3 from human melanoma cell line M21.

Application

Anti-Integrin aVβ3, clone LM609 ZooMAb, Cat. No. ZMS1030, is a recombinant Mouse monoclonal antibody that detects Integrin aVβ3 and is tested for use in Flow Cytometry, Immunocytochemistry, and Immunoprecipitation.
Immunoprecipitation Analysis: A representative lot of a non-recombinant antibody immunoprecipitated Integrin Vβ3 in Immunoprecipitation applications (Jessen, T.N., et. al. (2017). Exp Cell Res. 361(2):265-276).

Immunocytochemistry Analysis: A 1:100-1,000 dilution from a representative lot detected Integrin Vβ3 in HUVEC and MCF-7 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Integrin aVβ3 (UniProt: P06756/P05106; also known as CD51/CD61) is encoded by the ITGAV/ITGB3 (also known as MSK8, VNRA, VTNR/GP3A) genes (Gene ID: 3685/3690) in human. Integrin aV is a single-pass membrane protein that heterodimerizes with a beta subunit. The exact combination of various a and β subunits dictates the binding specificity of integrins to different ECM components. Although both subunits are required for adhesion, the binding specificity primarily depends on the extracellular region of the a subunit. aV associates with either β-1, β-3, β-5, β-6, or β-8 in a non-covalent manner. The structural and functional diversity of the integrin family are based upon the pairing abilities of the individual a and β subunits. Integrins are not constitutively active. Activation of an integrin from low ligand-binding affinity state to high ligand-binding affinity state requires conformational change that can originate from either from their cytoplasmic or extracellular interactions. The recognition site for most integrins that bind the ECM consists of an RGD (arginine-glycine-aspartic acid) sequence. Integrins bind to their ligands with low affinity and this binding occurs only when a certain minimum number of integrins are present at specific points known as focal contacts. Integrin Vβ3 serves as a receptor for cytotactin, fibronectin, laminin, matrix metalloproteinase-2, osteopontin, osteomodulin, prothrombin, thrombospondin, vitronectin and von Willebrand factor. Both Integrin aV and β3 are single-pass membrane proteins that are synthesized with a signal peptide (aa 1-30 and 1-26, respectively), which is subsequently cleaved off to generate the mature form. Their mature forms contain an extracellular domain (aa 31-992/27-718), a transmembrane domain (aa 993-1016/719-741), and a cytoplasmic domain (aa 1017-1048/742-788). This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service