Skip to Content
Merck
All Photos(1)

Key Documents

SAB5700525

Sigma-Aldrich

Anti-pan Phospho-Serine/Threonine antibody produced in rabbit

Synonym(s):

Anti-phosphorylation antibody, Phospho-serine antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€373.00

€373.00


Check Cart for Availability


Select a Size

Change View
100 μL
€373.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€373.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

concentration

0.38 mg/ml

technique(s)

western blot: 1:500-1:2000

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pan pSer/pThr)

Immunogen

A synthetic peptide corresponding to a sequence containing phosphorylated Serine/Threonine.

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. which secondary antibody fits with this primary antibody which is the "Anti-pan Phospho-Serine/Threonine antibody produced in rabbit"?

    1 answer
    1. The western blotting image at the top of the product page describes the secondary antibody used as HRP goat anti-rabbit IgG (H+L) at a dilution of 1:10,000 using ECL detection.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service