Skip to Content
Merck
All Photos(1)

Key Documents

Z759279

Greiner CELLSTAR® 384 well plates

flat bottom white polystyrene wells, sterile

Synonym(s):

384 multiwell plates, 384 well microplates, 384 well plates, Greiner tissue culture treated multiwell plates, 384-well

Sign Into View Organizational & Contract Pricing

Select a Size

32 EA
€260.00

€260.00


Check Cart for Availability


Select a Size

Change View
32 EA
€260.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
flat bottom white polystyrene wells
size:
384 wells
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid

€260.00


Check Cart for Availability

material

flat bottom white polystyrene wells

sterility

sterile

feature

lid

packaging

pack of 32 ea

manufacturer/tradename

Greiner 781080

maximum volume

131 μL

size

384 wells

working volume

110 μL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

GreinerCELLSTAR® 384 well plates feature flat bottom plates with rounded corners and white polystyrene wells. It is designed with improved cell adhesion via physical surface treatment making it suitable for cell culture applications.

Application

Greiner CELLSTAR® 384 well plates have been used for culturing HEK293T cells during TANGO assay to study the biological activity and specific targets of pollutants.[1]

Features and Benefits

  • DNase, RNase, human DNA free
  • Physical surface treated for better cell adhesion
  • Non-cytotoxic and non-pyrogenic
  • Sterile, stackable, and has a solid bottom
  • The lid enables minimal gas exchange due to low evaporation
  • Compatible with automated systems
  • Alphanumeric coding of well
  • Rounder square flat bottom well profile

Legal Information

CELLSTAR is a registered trademark of Greiner Bio-One GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua C Wilkinson et al.
Chemical research in toxicology, 37(2), 439-449 (2024-01-31)
Exposure to environmental pollutants is linked to numerous toxic outcomes, warranting concern about the effect of pollutants on human health. To assess the threat of pollutant exposure, it is essential to understand their biological activity. Unfortunately, gaps remain for many

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service