Documents
Z756954
Inhalation narcosis chamber
clear acrylic, pack of 1 ea
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
material
clear acrylic
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H46420-0002
General description
The Inhalation narcosis chamber has a space at the bottom filled with cotton soaked by anesthetic, and a vented floor directly above it to support the animal. Dimensions: I.D. is 197 mm (7 3/4 in.), top I.D. is 178 mm (7 in.), internal height is 127mm (5 in.), height below platform is 57 mm (2 1/4 in.), overall height is 197 mm (7 3/4 in.).
  • Optimally sized anesthetizing chamber for small animals including rats, mice, hamsters and guinea pigs
  • Body constructed of polypropylene and is unaffected by anesthesia chemicals
  • Features funnel for addition of anesthetic
  • Side-mounted stopcock lets fresh air enter to allow animal to breathe