Z742778
smartPans™ cahn style aluminim weighing dish
pack of 20 (Dishes supplied in flip top vial)
All Photos(1)
About This Item
Recommended Products
material
aluminum
packaging
pack of 20 (Dishes supplied in flip top vial)
manufacturer/tradename
LevGo Inc 41568-020
diam. × H × thickness
25 mm × 3.59 mm × 0.038 mm
weight
53 mgEach boat
General description
Ideal for microbalances, organic micro elementary / thermal analysis, and pharmaceutical use.
- Made of Ultra High Vacuum (UVH) Foil - “A” wettable and completely free from oils normally found on other grades of aluminum foil.
- Foil meets ASTM B-479, specification 3.1.4 and 10.3.1.
- “Cahn style” accessory pan / disc format
- Compatible with most commercially available analyzers and microbalances
- Foil and Pans are made in the USA
- Packaged 20 or 100 per easy-to-use PP flip top vials - Both the pans and the vials recyclable
Legal Information
smartPans is a trademark of LevGo Inc.