Skip to Content
Merck
All Photos(1)

Key Documents

Z742255

eXTReme FoilSeal Sealing Foils

for long-term compound storage, light sensitive assays and robotics, W × L 78.3 mm × 129 mm, non-sterile

Synonym(s):

aluminium plate seal, long term storage seal, robotic plate seal, universal multiplate seal

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€117.00

€117.00


Check Cart for Availability


Select a Size

Change View
100 EA
€117.00

About This Item

UNSPSC Code:
41122410
NACRES:
NC.01

€117.00


Check Cart for Availability

material

aluminum film

sterility

non-sterile

packaging

pkg of 100 ea

manufacturer/tradename

Excel Scientific XTR-FOIL-100

technique(s)

PCR: suitable

W × L

7.83 cm × 12.9 cm
78.3 mm × 129 mm

thickness

1.9 mil

General description

XTR foils offer high performance under extreme conditions. A 48 um soft non-permeable aluminum foil sealing film with 30 um layer of strong acrylic adhesive. FoilSeal sealing foils remove cleanly from plate surfaces with no adhesive stringing, residue or contamination. Each sealing foil measures 78.3 by 129 mm and offers sufficient sealing area for all plate types (e.g. flat top and raised-rim). Chamfered corners result in no plate overhang for robotics applications. Features one end-tab with perforations. Length between the perforation with end tab removed and the end of the foil seal is 118.1 mm.

Features and Benefits

  • Universal fit for use with all plate types (e.g. flat top and raised-rim)
  • The smaller dimensions and chamfered corners result in no plate overhang for robotics applications
  • Single end-tab features a custom perforation that will not tear during removal
  • Excellent barrier properties minimizes sample evaporation
  • 1.9mil thick aluminum foil minimizes shredding during removal and is ideal for cold storage
  • Conformable foil allows visual verification of seal security
  • Pierceable with pipet tips and robotic probes for direct sample retrieval
  • Specially formulated adhesive provides an excellent seal for all plate materials (e.g. polystyrene, polypropylene and two component polycarbonate/polypropylene PCR plates)
  • Foil will remove cleanly from plate surfaces with no adhesive stringing or residue

Legal Information

FoilSeal is a trademark of Excel Scientific, Inc.
eXTReme is a trademark of Excel Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service