Skip to Content
Merck
All Photos(1)

Key Documents

Z742249

Lena Biosciences SeedEZ 3D Cell Culture System

sheet size 5 cm × 10 cm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€152.00

€152.00


Check Cart for Availability


Select a Size

Change View
1 EA
€152.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.22

€152.00


Check Cart for Availability

sterility

sterile

packaging

sheet of 1 ea

manufacturer/tradename

Lena Biosciences SC-S510-0001

parameter

80 °C max. temp.

sheet size

5 cm × 10 cm

thickness

400 μm , dry state

color

white to off-white

pore size

~90 % porosity

Looking for similar products? Visit Product Comparison Guide

General description

SeedEZ is an absorbent 3D cell culture substrate suitable for culturing most cell types. It is compatible with most cell culture reagents, coatings, gels, and extracellular matrices. Please continue to use protocols and readouts you normally use. SeedEZ lets you reveal higher content information from cell cultures without sacrificing throughput.

SeedEZ provides for native tissue-like 3D cell environment, and in vivo like round cell morphology. This is vital for physiologically closer cell function, and cell network formation.

SeedEZ requires no special skills or prior experience with 3D cell cultures to homogeneously and reproducibly organize and culture cells in 3D. SeedEZ makes reproducible and consistent 3D cultures at seeding, which mature into reproducible and consistent tissue analogs for more conclusive and predictive results in life sciences research and drug discovery.

Other Notes

Use or cut to a desired shape and size with sterile scissors.

Legal Information

SeedEZ is a trademark of Lena Biosciences, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paula G Miller et al.
Biotechnology and bioengineering, 117(2), 486-497 (2019-10-15)
Efficient and economical delivery of pharmaceuticals to patients is critical for effective therapy. Here we describe a multiorgan (lung, liver, and breast cancer) microphysiological system ("Body-on-a-Chip") designed to mimic both inhalation therapy and/or intravenous therapy using curcumin as a model

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service