Skip to Content
Merck
All Photos(1)

Key Documents

Z742096

Western Blot Box

clear, size 7.3 cm × 3.0 cm × 1.9 cm, pk of 5

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
€24.60

€24.60


Check Cart for Availability


Select a Size

Change View
5 EA
€24.60

About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

€24.60


Check Cart for Availability

packaging

pk of 5

size

7.3 cm × 3.0 cm × 1.9 cm

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

  • Perfect box for every blot
  • Reduce antibody usage
  • Hinged Lid minimizes evaporation
  • For halves, wholes, strips, or minis


No matter what size your Western blot is, there′s a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at 90° angles and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies (for example, popular mini-gels require only about 4mL).
At a price that makes them truly disposable, all sizes are engineered from crystal clear polystyrene. All sizes are also equipped with an easy-to-open hinged lid that reduces evaporation.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service