Skip to Content
Merck
All Photos(1)

Documents

BAF451000002

Scienceware® gel incubating tray

W × L 16.5 cm (6.5 in.) × 16.5 cm (6.5 in.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.51

packaging

pack of 5 ea

manufacturer/tradename

Bel-Art F45100-0002

W × L

16.5 cm (6.5 in.) × 16.5 cm (6.5 in.)

Looking for similar products? Visit Product Comparison Guide

General description

Minimize the use of expensive antibodies when incubating filters in the Western Blot process by using an optimally sized tray.

  • Includes attached lid to protect against spillage
  • Trays are stackable, reclosable and reusable
  • Molded of polypropylene
  • Autoclavable at 121°C (250°F)

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service