Skip to Content
Merck
All Photos(1)

Key Documents

Z741970

Cameo Filtration Membrane

polycarbonate membrane (Track Etched (PCTE)), pore size 0.4 μm, diam. 47 mm, pkg of 100 ea

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€165.00

€165.00


Check Cart for Availability


Select a Size

Change View
100 EA
€165.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.24

€165.00


Check Cart for Availability

material

polycarbonate membrane (Track Etched (PCTE))

sterility

non-sterile

packaging

pkg of 100 ea

manufacturer/tradename

GVS Life Sciences 1215617

diam.

47 mm

pore size

0.4 μm

Looking for similar products? Visit Product Comparison Guide

General description

Designed to speed up and increase sample volume throughput while reducing back pressure, the hold-up volume is slightly lower due to improved flow channel design and reduced spacing between the supports. The design improvements allow an increase in operating pressure the syringe filters can withstand.

Applications: Analysis of Biological and Organic Fluids, Analytical Sample Prep, HPLC, Composite assays, Content uniformity, Dissolution testing, Environmental samples, Food Analysis

Legal Information

Cameo is a trademark of GVS S.p.A.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service