Skip to Content
Merck
All Photos(1)

Key Documents

Z740424

Tamper proof specimen containers

size 5700 mL

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
€152.00

€152.00


Check Cart for Availability


Select a Size

Change View
10 EA
€152.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

€152.00


Check Cart for Availability

material

polypropylene cap
polypropylene container

packaging

pack of 10 ea

manufacturer/tradename

Simport C581-5700

size

5700 mL

Looking for similar products? Visit Product Comparison Guide

General description

These disposable containers are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples. Tight-fitting lids prevent leaks and odors. They incorporate a tamperproof lid which is opened by first removing tab on side of lid, then by lifting the cover. They resist temperatures up to 80 °C. The two larger sizes are supplied with a plastic handle for easy carrying.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service