Skip to Content
Merck
All Photos(1)

Key Documents

Z273260

Sigma-Aldrich

Scienceware® Aquet® liquid detergent

pkg of 1 × 3.8 L (1 Gal)

Synonym(s):

cleabing concentrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€87.70

€87.70


Check Cart for Availability


Select a Size

Change View
1 EA
€87.70

About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

€87.70


Check Cart for Availability

description

non-ionic

packaging

pack of 1 ea
pkg of 1 × 3.8 L (1 Gal)

manufacturer/tradename

Bel-Art F17094-0030

technique(s)

HPLC: suitable
protein quantification: suitable

General description

Neutral pH wetting agent and detergent for laboratory glassware and optics. Diluted to 1% solution. Removes surface film and grease, permitting complete drainage of glassware. Biodegradable, phosphate-free, and nonionic.

Legal Information

Aquet is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

534.2 - 555.8 °F

Flash Point(C)

279 - 291 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chantelle Venter et al.
International journal of molecular sciences, 21(21) (2020-11-07)
Progressive respiratory failure is seen as a major cause of death in severe acute respiratory syndrome coronavirus 2 (SARS-Cov-2)-induced infection. Relatively little is known about the associated morphologic and molecular changes in the circulation of these patients. In particular, platelet

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service