Skip to Content
Merck
All Photos(1)

Documents

Z722502

X-Pierce film

non-sterile

Synonym(s):

piercable film, piercable plate seal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

material

vinyl film

sterility

non-sterile

feature

piercable

manufacturer/tradename

Excel Scientific XP-100

packaging

pkg of One tamper-evident bag of 100

technique(s)

tissue processing: suitable

W × L

79.4 mm × 145.5 mm

Looking for similar products? Visit Product Comparison Guide

General description

Also useful for protecting contents of plate from contamination during multiple manual reagent additions over time. X-Pierce films are 89 μm thick vinyl with a 18 μm thick adhesive layer, designed for temporary protection from contamination and evaporation for samples in 96 well plates. A precut "X" over each well creates four flaps that easily bend downward when pushed by a robotic probe or pipette tip, allowing sample access without coring or adhesive fouling. A thin adhesive layer and special hairline cuts separating the X-Pierce flaps combine to prevent probes from contacting adhesive and becoming occluded. The flaps return to their original position after sampling for continued protection. For long-term sample protection after sampling, a continuous film should be applied as a second layer.

Legal Information

X-Pierce is a trademark of Excel Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service