Skip to Content
Merck
All Photos(1)

Key Documents

Z369667

Sigma-Aldrich

SealPlate® film

sterile

Synonym(s):

ELISA plate seals, qPCR plate seal

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€104.00

€104.00


Check Cart for Availability


Select a Size

Change View
100 EA
€104.00

About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

€104.00


Check Cart for Availability

material

polyester film

sterility

sterile

feature

piercable: no

packaging

pack of 100 ea

manufacturer/tradename

Excel Scientific STR-SEAL-PLT

parameter

-40 to +120 °C temperature

technique(s)

ELISA: suitable

L

141 mm

W × L

7.94 cm × 14.1 cm
79.4 mm × 141 mm

W

79.4 mm

size

Up to 1536 wells

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

Sheets pre-cut to fit standard multiwell plates. Both film and adhesive are inert and compatible with microplate procedures. Adhesive forms a tight, waterproof seal, preventing cross-contamination and evaporation. SealPlate film is 2 mil (0.05 mm) polyester, with a functional temperature range of -40 to +120 °C.

Application

SealPlate® film is suitable for sealing tissue culture plates for temporary storage, incubation, and containment of biohazards. It can also be used in sensitive enzyme-linked immunosorbent assay (ELISA) for sealing of wells to avoid edge effect.

Legal Information

SealPlate is a registered trademark of Excel Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca Sternke-Hoffmann et al.
International journal of molecular sciences, 21(6) (2020-03-19)
The amyloid fibril formation by α -synuclein is a hallmark of various neurodegenerative disorders, most notably Parkinson's disease. Epigallocatechin gallate (EGCG) has been reported to be an efficient inhibitor of amyloid formation by numerous proteins, among them α -synuclein. Here
Tomas Sneideris et al.
Biomolecules, 9(12) (2019-12-15)
Millions of people around the world suffer from amyloid-related disorders, including Alzheimer's and Parkinson's diseases. Despite significant and sustained efforts, there are still no disease-modifying drugs available for the majority of amyloid-related disorders, and the overall failure rate in clinical

Questions

1–2 of 2 Questions  
  1. Is this gas permeable or impermeable?

    1 answer
    1. The SealPlate sealing film products are not gas permeable, but will prevent cross contamination. Should permeability be required, see the Aeraseal and SealMate products at the link below:
      https://www.sigmaaldrich.com/search/aeraseal?focus=products&page=1&perpage=30&sort=relevance&term=aeraseal&type=product

      Helpful?

  2. Does the film allow gas exchange? What is the water vapor transmission rate?

    1 answer
    1. This item is a product of Excel Scientific. This material does not allow for gas exchange. The vapor transmission rate has not been established. Note that the AeraSeal membranes do allow for gas exchange, see product Z721573 to review a sterile version.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service