Skip to Content
Merck
All Photos(1)

Documents

Z707430

AeraSeal film

non-sterile

Synonym(s):

ELISA plate seal, breathable plate seal, permeable rayon film, piercable plate seal, qPCR plate seal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

material

Rayon film

sterility

non-sterile

feature

piercable

packaging

pack of 100 ea

manufacturer/tradename

Excel Scientific B-100

technique(s)

tissue culture: suitable

L

142.9 mm

W × L

8.26 cm × 14.29 cm

W

82.6 mm

Looking for similar products? Visit Product Comparison Guide

General description

A 140 μm thick hydrophobic porous sealing film with medical-grade adhesive for tissue culture plates, bio-blocks, and 96-well plates where gas exchange is necessary for cell or bacterial growth. AeraSeal sealing films minimize cross-contamination, spillage and evaporation. They allow uniform air and CO2 exchange for all wells, unlike plate lids which favor exchange for wells near the plate edges. Dimensions: 82.6 by 142.9 mm for standard-size tissue culture plate
  • non-cytotoxic, highly gas permeable
  • easily pierceable with pipet tips or pipets for sample recovery
  • recommended for temperatures from -20 °C to +80 °C


Note: AeraSeal films are suitable for hypoxic culture conditions.

Legal Information

AeraSeal is a trademark of Excel Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service