Key Documents
Z659703
Bandelin SONOREX Ultrasonic bath accessories
tank lid for Z659673 and Z659029
Select a Size
1 EA
€59.00
€59.00
Estimated to ship on13 May 2025
Select a Size
Change View
1 EA
€59.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.74
€59.00
Estimated to ship on13 May 2025
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Bandelin D 255
availability
not available in USA and Canada
General description
Stainless steel lid for an ultrasonic bath protects the bath liquid from external contamination and prevents unpleasant odors from the cleaning or disinfection agents added. In addition, the noise emitted when the ultrasonic cleaner is in operation, which may be perceived as annoying, is reduced. The lids are designed in such a way that any condensation that occurs is directed back into the bath.
Features and Benefits
  • protection the bath fluid from contamination
  • sound-absorbing
  • shortens the heating phase
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)