Skip to Content
Merck
All Photos(1)

Key Documents

Z358207

Sigma-Aldrich

Nalgene® vacuum filtration system

filter capacity 1000 mL, pore size 0.2 μm

Synonym(s):

bottletop filtration system, sterile filtration

Sign Into View Organizational & Contract Pricing

Select a Size

1 CASE
€358.00

€358.00


Check Cart for Availability


Select a Size

Change View
1 CASE
€358.00

About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

€358.00


Check Cart for Availability

material

polyethersulfone membrane

sterility

sterile

feature

cap plug seal
graduations

packaging

pack of 12

manufacturer/tradename

Nalgene® 567-0020

bottle capacity

1000 mL

filter capacity

1000 mL

funnel capacity

1000 mL

membrane diam.

90 mm

pore size

0.2 μm

Looking for similar products? Visit Product Comparison Guide

General description

Increase filtration efficiency by using the Nalgene® Rapid-Flow disposable filter units featuring the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination when filtering cell culture media, serum, additives and buffers. With a graduated upper chamber and Polyethersulfone (PES) membrane this filter unit offer higher aqueous flow rates; lower extractables (no wetting agent); lower protein binding; and greater strength, capacity, and chemical resistance than cellulose acetate, nitrocellulose, or nylon.

Application

Nalgene® vacuum filtration system is suitable for the filtration of serum, cell culture medium, buffers and additives.

Features and Benefits

Nalgene® vacuum filtration system have the following benefits:
  • Polyethersulfone (PES) membrane is best suited for cells and tissue culture.
  • Faster results and less waiting time
  • Non-pyrogenic and endotoxin-free
  • Graduated upper chamber

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service