Key Documents
Z336823
BRAND® Imhoff cone rack
PMMA
Select a Size
1 EA
€119.00
€119.00
Check Cart for Availability
Select a Size
Change View
1 EA
€119.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.12
€119.00
Check Cart for Availability
material
PMMA
feature
Holds 2 x Imhoff cones
manufacturer/tradename
BRAND 388050
L × W × H
300 mm × 130 mm × 315 mm
General description
The BRAND® Imhoff Cone Rack is designed to securely hold two Imhoff sedimentation cones made of plastic, providing a practical solution for sedimentation processes. Constructed from a combination of PMMA and PP, this rack offers durability and stability while remaining lightweight. Its compact design makes it easy to carry, even when the cones are filled, facilitating convenient transport and handling in laboratory settings. This essential accessory enhances the efficiency of sedimentation experiments, ensuring that your cones are securely supported and easily accessible.
Other Notes
BR388050 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)