Key Documents
Z332348
Bottle adapters for BRAND® Dispensers and Burettes
GL 28 thread, GL 28 bottle (/S28), ETFE
Select a Size
1 EA
€14.60
€14.60
Estimated to ship on29 May 2025
Select a Size
Change View
1 EA
€14.60
About This Item
UNSPSC Code:
41121815
NACRES:
NB.01
€14.60
Estimated to ship on29 May 2025
material
ETFE
manufacturer/tradename
BRAND 704378
fitting
GL 28 bottle (/S28)
GL 28 thread
General description
For all Dispensette® models as well as seripettor® and QuikSip
Other Notes
BR704378 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)