Skip to Content
Merck
All Photos(1)

Key Documents

Z0173

Sigma-Aldrich

Zinc chloride

BioReagent, for molecular biology, ≥97.0%

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

Molecular Biology
for molecular biology

vapor pressure

1 mmHg ( 428 °C)

product line

BioReagent

Assay

≥97.0%

mp

293 °C (lit.)

solubility

H2O: soluble

cation traces

Fe: ≤0.001%
Mg: ≤0.01%
Pb: ≤0.005%

foreign activity

DNase and protease, none detected

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaushik Nath et al.
Environmental science and pollution research international, 20(6), 4030-4045 (2012-12-06)
Utilization of agrowaste materials for the production of activated carbon, as an excellent adsorbent with large surface area, is well established industrially, for dephenolation of wastewater. In the present work, dried pods of Prosopis cineraria-a novel and low-cost agrowaste material-were
Michele K Sankako et al.
Pharmacological reports : PR, 64(6), 1537-1546 (2013-02-15)
The aim of this study was to evaluate the changes in testicular function of rats due to cigarette smoke exposure and the possible mechanism by which zinc protects against these alterations. Male Wistar rats (60 days old) were randomly divided
Shigeru Hishinuma et al.
Journal of pharmacological sciences, 119(3), 233-242 (2012-07-13)
We examined the regulatory mechanisms of the affinity of Gq protein-coupled histamine H(1)-receptors for histamine after histamine pretreatment in intact human U373 MG astrocytoma cells. In control cells, the displacement curves for histamine against the binding of 5 nM [(3)H]mepyramine
Hatice Yorulmaz et al.
Biological trace element research, 151(2), 256-262 (2012-11-28)
We evaluated the effect of zinc treatment on the blood-brain barrier (BBB) permeability and the levels of zinc (Zn), natrium (Na), magnesium (Mg), and copper (Cu) in the brain tissue during epileptic seizures. The Wistar albino rats were divided into
L Buizza et al.
Cell death & disease, 4, e484-e484 (2013-02-09)
The role of p53 in neurodegenerative diseases is essentially associated with neuronal death. Recently an alternative point of view is emerging, as altered p53 conformation and impaired protein function have been found in fibroblasts and blood cells derived from Alzheimer's

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service