Skip to Content
Merck
All Photos(1)

Documents

Z0152

Sigma-Aldrich

Zinc chloride

BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture

Synonym(s):

Dichlorozinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71

grade

for molecular biology

Quality Level

vapor pressure

1 mmHg ( 428 °C)

product line

BioReagent

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

mp

293 °C (lit.)

solubility

H2O: may be turbid 20 g/200 ML + 6 mL 1M HCL, clear to slightly hazy, colorless to faint yellow or tan

foreign activity

DNAse, none detected
Endonuclease, none detected
Exonuclease, none detected
NICKase, none detected
Protease, none detected

SMILES string

[Cl-].[Cl-].[Zn++]

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karla Viridiana Castro-Cerritos et al.
Bio-protocol, 7(23), e2634-e2634 (2017-12-05)
Elucidating how a population of non-growing bacteria generates mutations improves our understanding of phenomena like antibiotic resistance, bacterial pathogenesis, genetic diversity and evolution. To evaluate mutations that occur in nutritionally stressed non-growing bacteria, we have employed the strain B. subtilis
Christopher J Harbort et al.
STAR protocols, 1(3), 100226-100226 (2020-12-31)
We present a gnotobiotic system for microbiota reconstitution on Arabidopsis thaliana under contrasting iron availability. This system induces iron starvation in plants by providing an unavailable form, mimicking conditions in alkaline soils. Inoculation of taxonomically diverse bacteria reconstitutes plants with
Siamak Redhai et al.
Nature, 580(7802), 263-268 (2020-04-10)
In cells, organs and whole organisms, nutrient sensing is key to maintaining homeostasis and adapting to a fluctuating environment1. In many animals, nutrient sensors are found within the enteroendocrine cells of the digestive system; however, less is known about nutrient
Manuella P Kaster et al.
Pharmacological reports : PR, 64(3), 706-713 (2012-07-21)
In this work, the contribution of NMDA receptors to the antidepressant-like effect of adenosine in the forced swimming test (FST) was investigated. The pretreatment of mice with NMDA or D-serine was able to prevent the anti-immobility effect of either adenosine
Andrea M Hui et al.
Dermatologic clinics, 30(3), 503-515 (2012-07-18)
For melanoma treatment, the primary goal is complete removal with histologically negative margins. Mohs micrographic surgery (MMS) has been extensively used and studied for the treatment of nonmelanoma skin cancer, particularly at sites where tissue conservation is vital. The use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service