Skip to Content
Merck
All Photos(4)

Key Documents

XNASS

Sigma-Aldrich

REDExtract-N-Amp Seed PCR Kit

sufficient for 10 extractions, sufficient for 10 amplifications

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€35.40

€35.40


Check Cart for Availability


Select a Size

Change View
1 KIT
€35.40

About This Item

UNSPSC Code:
41106303
NACRES:
NA.55

€35.40


Check Cart for Availability

usage

sufficient for 10 amplifications
sufficient for 10 extractions
sufficient for 10 reactions

feature

dNTPs included
hotstart

technique(s)

PCR: suitable

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The REDExtract-N-Amp Seed PCR Kits provide all the reagents necessary to rapidly extract DNA from a wide variety of plant seeds and amplify targets of interest by direct PCR (polymerase chain reaction). A novel extraction method eliminates the need for long enzymatic digestions. The kit also includes a specially formulated hot start PCR reaction mix for amplification directly from the extract. The REDExtract-N-Amp PCR ReadyMix contains an inert dye that acts as a tracking dye and allows for convenient loading of PCR reactions onto agarose gels for analysis.

Application

REDExtract-N-Amp seed PCR Kit has been used for DNA extraction[1][2]

Features and Benefits

  • Perfect for quick genotyping results
  • Rapid extraction of genomic DNA for PCR in 15 minutes
  • No long enzymatic digestions
  • Hot Start antibody for highly specific PCR amplification of genomic DNA
  • Compatible with a wide variety of seed sources
  • Extracts are stable at 4 °C for at least 6 months, allowing for multiple reassays
  • Phenol/chloroform extraction or alcohol precipitation not required
  • Column purification or centrifugation not required

Principle

The kit comes with validated protocols to extract and amplify genomic DNA from corn, wheat, sunflower, cotton, soybean, sorghum, canola, and Arabidopsis seeds. Additional protocols are available. In a typical procedure, genomic DNA is extracted from a sample that has been ground and incubated in the seed preparation solution and extraction solution for 10 minutes at room temperature. The sample is heated to 95 °C for 3 minutes and then mixed with a third solution to neutralize inhibitory substances prior to PCR. A portion of the DNA extract is then added to a PCR reaction containing primers and the Extract-N-Amp PCR Reaction Mix (included in the kit) and PCR is initiated.

Other Notes

For additional information, please see www.sigma-aldrich.com/extract-n-amp.

Legal Information

Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries. SYBR Green nucleic acid stain is licensed from Molecular Probes, Inc. under US Patent No. 5,436,134 for use in Real-Time PCR by end-users for research and development only. SYBR is a registered trademark of Molecular Probes, Inc.

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
REDExtract-N-Amp is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • Seed Preparation Solution

Kit Components Also Available Separately

Product No.
Description
SDS

  • E7526Extraction SolutionSDS

  • N3910Neutralization Solution BSDS

  • R4775REDExtract-N-Amp PCR Reaction MixSDS

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Marko Stojkovic et al.
Journal of separation science, 35(24), 3509-3513 (2012-12-12)
The use of CE with contactless conductivity detection for the determination of PCR products is demonstrated for the first time. The separation of specific length PCR products according to their size could be achieved using 5% PVP as a sieving
Angela Quarta et al.
FEMS microbiology letters, 259(1), 7-13 (2006-05-11)
The ability to rapidly distinguish trichothecene chemotypes in a given species/population of the genus Fusarium is important due to significant differences in the toxicity of these secondary metabolites. A multiplex PCR assay, based on primer pairs derived from the Tri3

Articles

Extract-N-Amp™ PCR kits are the world’s first integrated extraction and amplification process for rapid blood, tissue or plant assays.

Simple DNA/RNA purification methods aid genome analysis from various sources, enhancing research efficiency.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service