Key Documents
WPB01474
Whirl-Pak® Blender Bag
capacity 5441 mL (184 oz), bag W × L × thickness 37.5 cm × 50 cm × 0.102 mm
About This Item
UNSPSC Code:
24111500
NACRES:
NB.74
Pricing and availability is not currently available.
material
clear linear low-density polyethylene (LLDPE)
sterility
sterile
packaging
pkg of 500 ea (2 boxes of 250EA)
manufacturer/tradename
Nasco Whirl-pak® B01474
bag W × L × thickness
37.5 cm × 50 cm × 0.102 mm
capacity
5441 mL (184 oz)
General description
These bags are ideal for use in a homogenizer in the lab with all solid and liquid contents including but not limited to: food, protein, powder, pharmaceuticals, dairy, environmental and many more. Whirl-Pak® sterilized sampling bags are ideal for food & beverage, dairy, water, medical, veterinary, environmental, soil, and industrial markets as well as surface, forensic science, genetics, biomedical, and pharmaceutical sampling. Whirl-Pak® bags have been the professionals’ trusted sampling bag for over 60 years and offer puncture-proof closure tabs, guaranteed sterility, easy identification, and a leak-proof closure you can rely on.
Features and Benefits
  • Perforated for easy opening and to protect sterility
  • Designed for quick testing and disposal when the sample does not have to be retained
  • Bag can be frozen to any temperature, even with liquid nitrogen, but should not be used at temperatures above 180°F (82°C)
  • FDA and USDA Approved
  • Post-manufacturing sterilization
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)