Skip to Content
Merck
All Photos(2)

Key Documents

W104

Sigma-Aldrich

WIN 62,577

solid

Synonym(s):

17-β-Hydroxy-17-α-ethynyl-δ-4-androstano[3,2-b]pyrimido[1,2-a]benzimidazole

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€201.00
25 MG
€635.00

€201.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€201.00
25 MG
€635.00

About This Item

Empirical Formula (Hill Notation):
C29H31N3O
CAS Number:
Molecular Weight:
437.58
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€201.00


Estimated to ship on25 May 2025


Request a Bulk Order

form

solid

color

yellow

solubility

ethanol: 1.4 mg/mL
DMSO: 4 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

[H][C@@]12CCC3=Cc4nc5nc6ccccc6n5cc4C[C@]3(C)[C@@]1([H])CC[C@@]7(C)[C@@]2([H])CC[C@@]7(O)C#C

InChI

1S/C29H31N3O/c1-4-29(33)14-12-22-20-10-9-19-15-24-18(16-27(19,2)21(20)11-13-28(22,29)3)17-32-25-8-6-5-7-23(25)30-26(32)31-24/h1,5-8,15,17,20-22,33H,9-14,16H2,2-3H3/t20-,21+,22+,27+,28+,29+/m1/s1

InChI key

YBCYJDNOPAFFOW-XOAARHKISA-N

Gene Information

human ... TACR1(6869)
rat ... Tacr1(24807)

Biochem/physiol Actions

Non-peptide NK1 tachykinin receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B R Venepalli et al.
Journal of medicinal chemistry, 35(2), 374-378 (1992-01-24)
Several heterosteroids containing a dihydroethisterone skeleton were prepared and shown to displace substance P in a receptor binding assay. Further biochemical (kinetic and Scatchard analyses) and pharmacological evaluation (substance P-induced plasma extravasation and salivation in the rat) of a representative
K C Appell et al.
Molecular pharmacology, 41(4), 772-778 (1992-04-01)
125I-Bolton-Hunter-substance P (125I-BH-SP) binding properties of three novel classes of neurokinin-1 (NK-1) receptor antagonists were investigated in tissues derived from humans, guinea pigs, and rats. 125I-BH-SP was shown to bind to a single class of binding sites, with similar dissociation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service