Skip to Content
Merck
All Photos(1)

Key Documents

L0376

Sigma-Aldrich

L-Leucine β-naphthylamide hydrochloride

leucine aminopeptidase substrate

Synonym(s):

L-Leucine-2-naphthylamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€278.00
5 G
€473.00

€278.00


Estimated to ship on28 May 2025



Select a Size

Change View
1 G
€278.00
5 G
€473.00

About This Item

Empirical Formula (Hill Notation):
C16H20N2O · HCl
CAS Number:
Molecular Weight:
292.80
Beilstein:
3918167
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.32

€278.00


Estimated to ship on28 May 2025


Assay

≥98% (TLC)

form

powder

solubility

H2O: soluble

storage temp.

−20°C

SMILES string

Cl[H].CC(C)C[C@H](N)C(=O)Nc1ccc2ccccc2c1

InChI

1S/C16H20N2O.ClH/c1-11(2)9-15(17)16(19)18-14-8-7-12-5-3-4-6-13(12)10-14;/h3-8,10-11,15H,9,17H2,1-2H3,(H,18,19);1H/t15-;/m0./s1

InChI key

HTHKKWZMEVJWQF-RSAXXLAASA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Substrate for leucine aminopeptidase determination in colorimetric and histochemical procedures.

Quality

Very low free β-naphthylamine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Niinobe et al.
Journal of biochemistry, 87(1), 195-203 (1980-01-01)
Aminopeptidase and arylamidase from human liver were purified 1,580- and 1,130-fold, respectively, by ammonium sulfate fractionation, TEAE-cellulose and hydroxylapatite column chromatographies, and Sephadex G-200 gel filtration. The purified preparations appeared to be homogeneous on polyacrylamide gel electrophoresis. The molecular weights

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service