Skip to Content
Merck
All Photos(1)

Key Documents

W0270

Sigma-Aldrich

WAY 161503 hydrochloride

≥98% (HPLC)

Synonym(s):

8,9-Dichloro-2,3,4,4a-tetrahydro-1H-pyrazino[1,2-a]quinoxalin-5(6H)-one hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€116.00

€116.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€116.00

About This Item

Empirical Formula (Hill Notation):
C11H11Cl2N3O · HCl
CAS Number:
Molecular Weight:
308.59
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

€116.00


Estimated to ship on22 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: >10 mg/mL

originator

Wyeth

storage temp.

room temp

SMILES string

Cl.Clc1cc2NC(=O)C3CNCCN3c2cc1Cl

InChI

1S/C11H11Cl2N3O.ClH/c12-6-3-8-9(4-7(6)13)16-2-1-14-5-10(16)11(17)15-8;/h3-4,10,14H,1-2,5H2,(H,15,17);1H

InChI key

YPNWSZJDAKOUAW-UHFFFAOYSA-N

Biochem/physiol Actions

The 5-HT2C receptors have been implicated in conditions including obesity, anxiety, depression, OCD, schizophrenia, migraine, nociception and erectile dysfunction.

Features and Benefits

This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam L Halberstadt et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 34(8), 1958-1967 (2009-03-27)
Although it is well established that hallucinogens act as 5-HT(2A) and 5-HT(2C) receptor agonists, little is known about the relative contributions of 5-HT(2A) and 5-HT(2C) receptors to the acute behavioral effects of these drugs. The behavioral pattern monitor was used

Articles

We offer many products related to serotonin receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service