Skip to Content
Merck

Skip To

SML2237

Ormeloxifene

≥95% (HPLC), SERM modulator, film or powder or oil

Synonym(s):

(trans)-1-[2-[p-(7-methoxy-2,2-dimethyl-3-phenyl-4-chromanyl)phenoxy]ethyl]pyrrolidine, 3,4-trans-2,2-Dimethyl-3-phenyl-4-[p-(β-pyrrolidinoethoxy)phenyl]-7-methoxychroman, Centchroman, rel-1-[2-[4-[(3R,4R)-3,4-Dihydro-7-methoxy-2,2-dimethyl-3-phenyl-2H-1-benzopyran-4-yl]phenoxy]ethyl]pyrrolidine, trans-Centchroman

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 102,00
€ 86,70
25 mg
Check Cart for Availability
€ 414,00
€ 351,90

About This Item

Empirical Formula (Hill Notation):
C30H35NO3
CAS Number:
Molecular Weight:
457.60
NACRES:
NA.77
UNSPSC Code:
12352200
MDL number:
Assay:
≥95% (HPLC)
Form:
film or powder (or oil)

€ 86,70

List Price€ 102,00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ormeloxifene, ≥95% (HPLC)

assay

≥95% (HPLC)

form

film or powder (or oil)

color

colorless to yellow-white

storage temp.

2-8°C

SMILES string

CC([C@@H](C1=CC=CC=C1)[C@@H]2C3=CC=C(OCCN4CCCC4)C=C3)(C)OC5=C2C=CC(OC)=C5

InChI

1S/C30H35NO3/c1-30(2)29(23-9-5-4-6-10-23)28(26-16-15-25(32-3)21-27(26)34-30)22-11-13-24(14-12-22)33-20-19-31-17-7-8-18-31/h4-6,9-16,21,28-29H,7-8,17-20H2,1-3H3/t28-,29+/m1/s1

InChI key

XZEUAXYWNKYKPL-WDYNHAJCSA-N

Biochem/physiol Actions

Ormeloxifene (Centchroman) is a non-steroidal selective estrogen receptor modulator (SERM) that exhibit anticancer activity. Ormeloxifene suppresses the ovariectomy-induced bone resorption in rats.
non-steroidal selective estrogen receptor modulator (SERM)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H6278H7904SML2607
description

≥95% (HPLC)

description

≥70% Z isomer (remainder primarily E-isomer)

description

≥98% Z isomer

description

≥98% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

film or powder (or oil)

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

color

colorless to yellow-white

color

-

color

-

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters