Skip to Content
Merck
All Photos(1)

Documents

V4390

Sigma-Aldrich

VUF 8430 dihydrobromide

≥97% (NMR)

Synonym(s):

2-[(Aminoiminomethyl)amino]ethyl carbamimidothioic acid ester, S-(2-guanidylethyl)-isothiourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11N5S· 2HBr
CAS Number:
Molecular Weight:
323.05
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (NMR)

form

solid

storage condition

desiccated

solubility

deionized water: 24 mg/mL

storage temp.

room temp

SMILES string

Br.Br.NC(=N)NCCSC(N)=N

InChI

1S/C4H11N5S.2BrH/c5-3(6)9-1-2-10-4(7)8;;/h1-2H2,(H3,7,8)(H4,5,6,9);2*1H

InChI key

GPWJSTKHQMIXCA-UHFFFAOYSA-N

Biochem/physiol Actions

VUF 8430 dihydrobromide is a potent histamine H4 receptor agonist, with a 33-fold selectivity over the histamine H3 receptor and negligible affinity for the other histamine receptor.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamara E Galarza et al.
Biochemical pharmacology, 180, 114177-114177 (2020-07-30)
Epithelial-mesenchymal transition (EMT) contributes to cell invasion and metastasis during the progression of epithelial cancers. Though preclinical evidence suggests a role for histamine H4 receptor (H4R) in breast cancer growth, its function in the EMT is less known. In this

Articles

We offers many products related to histamine receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service