Skip to Content
Merck

Skip To

C8138

Chlorpromazine hydrochloride

≥98% (TLC), powder, D2 dopamine receptor antagonist

Synonym(s):

2-Chloro-10-(3-dimethylaminopropyl)phenothiazine hydrochloride, CPZ, Largactil

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Check Cart for Availability
€ 157,00
25 g
Check Cart for Availability
€ 452,00
100 g
Check Cart for Availability
€ 1.050,00

About This Item

Empirical Formula (Hill Notation):
C17H19ClN2S · HCl
CAS Number:
Molecular Weight:
355.33
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
200-701-3
MDL number:
Beilstein/REAXYS Number:
3779989
Assay:
≥98% (TLC)
Form:
powder

€ 157,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Chlorpromazine hydrochloride, ≥98% (TLC)

Quality Segment

assay

≥98% (TLC)

form

powder

mp

194-196 °C

solubility

water: 50 mg/mL

originator

GlaxoSmithKline

SMILES string

Cl[H].CN(C)CCCN1c2ccccc2Sc3ccc(Cl)cc13

InChI

1S/C17H19ClN2S.ClH/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)21-17-9-8-13(18)12-15(17)20;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H

InChI key

FBSMERQALIEGJT-UHFFFAOYSA-N

Gene Information

Application

Chlorpromazine hydrochloride has been used:
  • as an antagonists for calmodulin kinase[1]
  • as a medium supplement to test its cytotoxic effects in human lung microvascular endothelial cells (HMVEC-L)
  • as an inhibitor of clathrin-mediated endocytosis in RAW 264.7 cells

Substitute for benzidine, o-dianisidine, and o-tolidine in the determination of microquantities of hemoglobin and peroxidase.

Biochem/physiol Actions

Chlorpromazine demonstrates cytotoxic and antiproliferative activity against leukemic cells, but does not affect the viability of normal lymphocytes.
Phenothiazine antipsychotic; D2 dopamine receptor antagonist, H1 histamine receptor antagonist; inhibits calmodulin-dependent stimulation of cyclic nucleotide phosphodiesterase and nitric oxide synthase.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
31679C0982BP856
form

powder

form

-

form

solid

form

solid

assay

≥98% (TLC)

assay

-

assay

98.0-101.5% dry basis

assay

-

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

-

solubility

water: 50 mg/mL

solubility

-

solubility

water: soluble 50 g/L

solubility

-

originator

GlaxoSmithKline

originator

-

originator

-

originator

-

Gene Information

human ... DRD2(1813), DRD3(1814), DRD4(1815), HRH1(3269), HTR2A(3356)

Gene Information

human ... DRD2(1813), DRD3(1814), DRD4(1815), HTR2A(3356)

Gene Information

human ... DRD2(1813), DRD3(1814), DRD4(1815), HTR2A(3356)

Gene Information

human ... DRD2(1813), DRD3(1814), DRD4(1815), HTR2A(3356)


Still not finding the right product?

Explore all of our products under Chlorpromazine hydrochloride


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Dear, I bought your product C8138 Chlorpromazine. I have few practical questions: - Can it be dissolved in DMSO? - Can it be dissolved in Phosphate-buffered saline? - What is the storage temperature of the dissolved product?

    1 answer
    1. This product is tested for solubility in water at 50 mg/mL. Other published sources state that this compound is also soluble in organic solvents such as DMSO and alcohols. Solution stability studies have not been perofrmed, however the Chlorpromazine HCl analytical standard, product C-904, is a 1 mg/mL solution in methanol. This item is not assigned an expiration date and is considered stable for at least 1 year from the date of shipment when stored at ?20°C.

      Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/966/223/c8138pis.pdf

      Please see the link below to review a publication related to the stability of this compound in water:
      http://onlinelibrary.wiley.com/doi/10.1038/sj.bjp.0705283/pdf

      Helpful?

Reviews

No rating value

Active Filters